Please login to your online account to display your discounted pricing

anti-XAF1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056355SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056355T

  • / Each
 

Description & Specifications

Specifications

Antigen XAF1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen Amino acids 201-218 (NQTSTMEKDVRPKTRSIN) and 301-317 (QLDLEKERYYKFKRFHF) of human XAF1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 54739
Test Specificity In human liver, a 34 kDa band should be observed.
Target Species Human
Target Species Murine