Please login to your online account to display your discounted pricing

anti-XAF1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056355

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056355

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54739
Test Specificity In human liver, a 34 kDa band should be observed.
Concentration 0.5mg/mL
Immunogen Amino acids 201-218 (NQTSTMEKDVRPKTRSIN) and 301-317 (QLDLEKERYYKFKRFHF) of human XAF1
Format Purified
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Antigen XAF1
Host Species Rabbit