Please login to your online account to display your discounted pricing

anti-WNVM, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus glycoprotein M. Genbank accession no. NP_776013.
Antigen WNVM
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056743

 
novus biologicals canada
NB10056743
Unlabeled 0.1mg Each for $329.00

NB10056743T

 
novus biologicals canada
NB10056743SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus glycoprotein M. Genbank accession no. NP_776013.
Antigen WNVM
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity West Nile Virus glycoprotein M (Genbank accession no. NP_776013).
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL