missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Wnt-5a, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 190-230 of human Wnt5A was used as the immunogen for this antibody
Antigen Wnt-5a
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224752 Novus Biologicals Canada
NBP224752
0.1mg Each for $362.00
NBP224752SS Novus Biologicals Canada
NBP224752SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
A synthetic peptide corresponding to amino acids 190-230 of human Wnt5A was used as the immunogen for this antibody
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
7474
0.5mg/mL
IgG
Wnt-5a
Purified
RUO
The amino acid sequence used as immunogen for the Wnt5A antibody is 100% homologous in human, cow, monkey, rat and mouse, 94% in opossum, 88% in xenopus, 81% in chimpanzee and 76% homologous in pufferfish.
100μg antibody in 200μL PBS containing 0.05% BSA