Please login to your online account to display your discounted pricing

anti-Wnt-5a, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224752

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224752

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Immunogen A synthetic peptide corresponding to amino acids 190-230 of human Wnt5A was used as the immunogen for this antibody
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen Wnt-5a
Gene ID (Entrez) 7474
Conjugate Unlabeled
Host Species Rabbit
Test Specificity The amino acid sequence used as immunogen for the Wnt5A antibody is 100% homologous in human, cow, monkey, rat and mouse, 94% in opossum, 88% in xenopus, 81% in chimpanzee and 76% homologous in pufferfish.
Regulatory Status RUO
Format Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg