Please login to your online account to display your discounted pricing

Wnt-4 Rat anti-Mouse, Clone: 55010, R&D Systems

Rat Monoclonal Antibody

$202.89 - $432.57

Specifications

Antigen Wnt-4
Clone 55010
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB475500

 
r & d systems
RDSMAB475500
Unlabeled 500μg Each for $432.57

MAB475050

 
r & d systems
RDSMAB475050
Unlabeled 50μg Each for $202.89
Description & Specifications

Specifications

Antigen Wnt-4
Clone 55010
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen E.Coli-derived recombinant mouse Wnt-4. aa 37-76 and 222-295
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Wnt-4 Monoclonal antibody MAB475 Specifically detects Wnt-4 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting