Please login to your online account to display your discounted pricing

Wnt-3a Rat anti-Human, Mouse, Clone: 217804, R&D Systems


Rat Monoclonal Antibody

Manufacturer: r & d systems  RDSMAB1324050

 View more versions of this product

Catalog No. MAB1324050

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Quantity 50μg
Isotype IgG2a
Clone 217804
Antigen Wnt-3a
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rat
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Immunogen E.Coli-derived recombinant mouse Wnt‑3a. Ser36-Gln75 and Trp219-Arg269
Purification Method Protein A or G purified from hybridoma culture supernatant

Wnt-3a Monoclonal antibody MAB1324SP Specifically detects Wnt-3a in Human, Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization, Western Blotting