Please login to your online account to display your discounted pricing

Wnt-3a Rat anti-Human, Mouse, Clone: 217804, R&D Systems

Rat Monoclonal Antibody

$207.26 - $441.90

Specifications

Antigen Wnt-3a
Clone 217804
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1324050

 
r & d systems
RDSMAB1324050
Unlabeled 50μg Each for $207.26

MAB1324500

 
r & d systems
RDSMAB1324500
Unlabeled 500μg Each for $441.90
Description & Specifications

Specifications

Antigen Wnt-3a
Clone 217804
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen E.Coli-derived recombinant mouse Wnt‑3a. Ser36-Gln75 and Trp219-Arg269
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Wnt-3a Monoclonal antibody MAB1324 Specifically detects Wnt-3a in Human, Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization, Western Blotting