Please login to your online account to display your discounted pricing

Wnt-3a Rat anti-Human, Mouse, Clone: 217804, R&D Systems


Rat Monoclonal Antibody

Manufacturer: r & d systems  RDSMAB1324500

 View more versions of this product

Catalog No. MAB1324500

  • / Each
 

Description & Specifications

Specifications

Immunogen E.Coli-derived recombinant mouse Wnt‑3a. Ser36-Gln75 and Trp219-Arg269
Isotype IgG2a
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Wnt-3a
Format Purified
Host Species Rat
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500μg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 217804
Conjugate Unlabeled

Wnt-3a Monoclonal antibody MAB1324500 Specifically detects Wnt-3a in Human, Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization, Western Blotting