Please login to your online account to display your discounted pricing

anti-WNK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600225SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600225SS

  • / Each
 

Description & Specifications

Specifications

Antigen WNK1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Gene Accession No. Q9H4A3
Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region of human WNK1 (within the last 350 amino acids). [UniProt No. Q9H4A3]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 65125
Target Species Human
Target Species Murine
Target Species Rat