Please login to your online account to display your discounted pricing

anti-WNK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600225

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600225

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 5mg/mL
Conjugate Unlabeled
Formulation PBS
Regulatory Status RUO
Gene ID (Entrez) 65125
Antigen WNK1
Gene Accession No. Q9H4A3
Primary or Secondary Primary
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide within the C-terminal region of human WNK1 (within the last 350 amino acids). [UniProt No. Q9H4A3]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL
Host Species Rabbit