Please login to your online account to display your discounted pricing

anti-WNK1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region of human WNK1 (within the last 350 amino acids). [UniProt No. Q9H4A3]
Antigen WNK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600225

 
novus biologicals canada
NB600225
Unlabeled 0.1mL Each for $309.05

NB600225SS

 
novus biologicals canada
NB600225SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region of human WNK1 (within the last 350 amino acids). [UniProt No. Q9H4A3]
Antigen WNK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q9H4A3
Gene ID (Entrez) 65125
Formulation PBS
Concentration 5mg/mL