Please login to your online account to display your discounted pricing

anti-WFS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001918SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001918SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. O76024
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.025mL
Gene ID (Entrez) 7466
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Regulatory Status RUO
Immunogen A 110 amino acid sequence from the N-terminal region of the human WFS1 protein. [UniProt No. O76024]
Antigen WFS1
Host Species Rabbit