Please login to your online account to display your discounted pricing

anti-WFS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001918

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001918

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A 110 amino acid sequence from the N-terminal region of the human WFS1 protein. [UniProt No. O76024]
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen WFS1
Gene Accession No. O76024
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7466
Host Species Rabbit
Format Antiserum
Regulatory Status RUO