Please login to your online account to display your discounted pricing

anti-WFS1, Polyclonal, Novus Biologicals™

$329.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A 110 amino acid sequence from the N-terminal region of the human WFS1 protein. [UniProt No. O76024]
Antigen WFS1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001918

 
novus biologicals canada
NB1001918
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A 110 amino acid sequence from the N-terminal region of the human WFS1 protein. [UniProt No. O76024]
Antigen WFS1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. O76024
Gene ID (Entrez) 7466