missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-West Nile Virus Protein E, Polyclonal, Novus Biologicals™

$139.00 - $373.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to 73-86 (CPTMGEAHNEKRAD) of West Nile virus envelope glycoprotein (NC_001563).
Antigen West Nile Virus Protein E
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056744 Novus Biologicals Canada
NB10056744
0.1mg Each for $373.00
NB10056744T Novus Biologicals Canada
NB10056744SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
West Nile Virus Protein E
Purified
RUO
PBS containing 0.05% BSA
A synthetic peptide corresponding to 73-86 (CPTMGEAHNEKRAD) of West Nile virus envelope glycoprotein (NC_001563).
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
0.5mg/mL