Please login to your online account to display your discounted pricing

anti-WDR24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP220892SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP220892SS

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 357 and 685 of WDR24 (Uniprot ID No. Q96S15)
Quantity 0.02mL
Antigen WDR24
Cross Reactivity Human
Concentration 0.96mg/mL
Conjugate Unlabeled
Formulation 1X PBS, 1% BSA, 20% Glycerol (pH 7).
Gene Accession No. Q96S15
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 84219
Format Affinity Purified