Please login to your online account to display your discounted pricing

anti-VprBP, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 935-985 from human VPRBP was used as the immunogen for this antibody.
Antigen VprBP
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227203SS

 
novus biologicals canada
NBP227203SS
Unlabeled 0.025mg Each for $109.00

NBP227203

 
novus biologicals canada
NBP227203
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 935-985 from human VPRBP was used as the immunogen for this antibody.
Antigen VprBP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, european pole-cat, pig, orangutan, rabbit, white-cheeked gibbon, opossum, wild turkey, african bush elephant, chicken, horse, Chinese hamster, guinea pig, and panda, 94% in cow, and platypus, and 88% homolgous in western clawed frog.There are numerous isoforms including 1505, 1504, 1076 and 783 amino acids.
Gene Accession No. Q9Y4B6
Gene ID (Entrez) 9730
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL