Please login to your online account to display your discounted pricing

anti-von Willebrand Factor (HRP), Polyclonal, Invitrogen™


Manufacturer: pierce biotechnology inc  PA174053

Catalog No. PIPA174053

  • / Each
 

Description & Specifications

Specifications

Antigen von Willebrand Factor (HRP)
Host Species Goat
Isotype IgG
Quantity 150μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Format Purified
Immunogen von Willebrand factor from human plasma.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate HRP