Please login to your online account to display your discounted pricing

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide common to both human and mouse Von Hippel Lindau protein.
Antigen Von Hippel Lindau
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100485SS

 
novus biologicals canada
NB100485SS
Unlabeled 0.025mL Each for $109.00

NB100485

 
novus biologicals canada
NB100485
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide common to both human and mouse Von Hippel Lindau protein.
Antigen Von Hippel Lindau
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. P40337
Gene ID (Entrez) 7428
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL