Please login to your online account to display your discounted pricing

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100485SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100485SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. P40337
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide common to both human and mouse Von Hippel Lindau protein.
Format Affinity Purified
Concentration 1mg/mL
Gene ID (Entrez) 7428
Primary or Secondary Primary
Quantity 0.025mL
Regulatory Status RUO
Host Species Rabbit
Cross Reactivity Human
Antigen Von Hippel Lindau