Please login to your online account to display your discounted pricing

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100485

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100485

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation Tris-citrate/phosphate, pH 7-8
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL
Format Affinity Purified
Host Species Rabbit
Antigen Von Hippel Lindau
Gene ID (Entrez) 7428
Gene Accession No. P40337
Conjugate Unlabeled
Concentration 1mg/mL
Immunogen A synthetic peptide common to both human and mouse Von Hippel Lindau protein.
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human