Please login to your online account to display your discounted pricing

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001899SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001899SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P40337
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Concentration 1mg/mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Antigen Von Hippel Lindau
Test Specificity Human. Not expected to work in mouse based on sequence divergence.
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7428
Format Affinity Purified
Conjugate Unlabeled
Cross Reactivity Human
Immunogen A synthetic peptide derived from the human Von Hippel Lindau protein. [UniProt No. P40337]
Regulatory Status RUO
Host Species Rabbit