Please login to your online account to display your discounted pricing

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001899

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001899

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL
Gene ID (Entrez) 7428
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. P40337
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Test Specificity Human. Not expected to work in mouse based on sequence divergence.
Format Affinity Purified
Immunogen A synthetic peptide derived from the human Von Hippel Lindau protein. [UniProt No. P40337]
Regulatory Status RUO
Antigen Von Hippel Lindau
Cross Reactivity Human
Quantity 0.1mL