Please login to your online account to display your discounted pricing

anti-visinin-like 1, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Chicken
Immunogen Recombinant full length His-tagged visinin-like 1 purified from E. coli.
Antigen visinin-like 1
Monoclonal or Polyclonal Polyclonal
Format Precipitation
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225168SS

 
novus biologicals canada
NBP225168SS
Unlabeled 0.025mL Each for $109.00

NBP225168

 
novus biologicals canada
NBP225168
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Chicken
Immunogen Recombinant full length His-tagged visinin-like 1 purified from E. coli.
Antigen visinin-like 1
Monoclonal or Polyclonal Polyclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, Bovine, Mouse and Rat. Other species have not been tested.
Gene Accession No. P62760
Gene ID (Entrez) 7447