Please login to your online account to display your discounted pricing

anti-Vimentin, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Recombinant full length purified Vimentin from E. coli
Antigen Vimentin
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225165

 
novus biologicals canada
NBP225165
Unlabeled 0.1mL Each for $309.05

NBP225165SS

 
novus biologicals canada
NBP225165SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant full length purified Vimentin from E. coli
Antigen Vimentin
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
Gene Accession No. P08670
Gene ID (Entrez) 7431