Please login to your online account to display your discounted pricing

anti-Vimentin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225165SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225165SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Format Antiserum
Regulatory Status RUO
Immunogen Recombinant full length purified Vimentin from E. coli
Conjugate Unlabeled
Gene Accession No. P08670
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Primary or Secondary Primary
Gene ID (Entrez) 7431
Host Species Rabbit
Antigen Vimentin
Quantity 0.025mL