Please login to your online account to display your discounted pricing

anti-Vimentin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225165

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225165

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. P08670
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Immunogen Recombinant full length purified Vimentin from E. coli
Regulatory Status RUO
Gene ID (Entrez) 7431
Quantity 0.1mL
Format Antiserum
Antigen Vimentin
Host Species Rabbit