missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-VGLL2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human VGLL2/VITO1 was used as the immunogen.
Antigen VGLL2
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224649 Novus Biologicals Canada
NBP224649
0.1mg Each for $255.80
NBP224649SS Novus Biologicals Canada
NBP224649SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 50-100 of human VGLL2/VITO1 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q8N8G2
100μg in 200μL PBS containing 0.05% BSA
IgG
VGLL2
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, horse and rat, 95% homologous in cow, 90% homologous in monkey and 85% homologous in mouse and dog.
245806
0.5mg/mL