Please login to your online account to display your discounted pricing

anti-VGLL2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224649SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224649SS

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Gene Accession No. Q8N8G2
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, horse and rat, 95% homologous in cow, 90% homologous in monkey and 85% homologous in mouse and dog.
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Quantity 0.025mg
Conjugate Unlabeled
Immunogen A portion of amino acids 50-100 of human VGLL2/VITO1 was used as the immunogen.
Gene ID (Entrez) 245806
Applications Western Blotting
Regulatory Status RUO
Antigen VGLL2
Format Purified