Please login to your online account to display your discounted pricing

anti-VEPH1, Polyclonal, Novus Biologicals™

$354.00 - $354.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen VEPH1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

38120002

 
novus biologicals canada
38120002
Unlabeled 0.05mg Each for $354.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen VEPH1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human VEPH1.
Gene Accession No. Q14D04
Gene ID (Entrez) 79674
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0