Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Residues corresponding to the kinase insert or C-terminus of mouse VEGF Receptor 2 (residues 1158-1365). [Swiss-Prot No. P35918]
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100627

 
novus biologicals canada
NB100627
Unlabeled 0.1mL Each for $329.00

NB100627SS

 
novus biologicals canada
NB100627SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Residues corresponding to the kinase insert or C-terminus of mouse VEGF Receptor 2 (residues 1158-1365). [Swiss-Prot No. P35918]
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene ID (Entrez) 3791