Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse VEGF Receptor 2 protein (between residues 1300-1367). [Swiss-Prot No. P35918]
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002382

 
novus biologicals canada
NB1002382
Unlabeled 0.1mL Each for $309.05

NB1002382SS

 
novus biologicals canada
NB1002382SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse VEGF Receptor 2 protein (between residues 1300-1367). [Swiss-Prot No. P35918]
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity This has only been shown to recognize the native form (180-190 kDa protein) of VEGFR-2, known as Flk-1 / KDR / VEGFR 2 (Vascular Endothelial Growth Factor Receptor 2), though it was raised against the precursor form. Human. Predicted to react with mouse based on 100% sequence homology. Immunogen sequence has 79% homology to rat. Hamster reactivity reported in scientific literature (PMID: 23763710).
Gene ID (Entrez) 3791
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL