Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100627SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100627SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 3791
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Immunogen Residues corresponding to the kinase insert or C-terminus of mouse VEGF Receptor 2 (residues 1158-1365). [Swiss-Prot No. P35918]
Primary or Secondary Primary
Host Species Rabbit
Format Antiserum
Quantity 0.025mL
Antigen VEGF R2/KDR/Flk-1