Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002382SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002382SS

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse VEGF Receptor 2 protein (between residues 1300-1367). [Swiss-Prot No. P35918]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3791
Test Specificity This has only been shown to recognize the native form (180-190 kDa protein) of VEGFR-2, known as Flk-1 / KDR / VEGFR 2 (Vascular Endothelial Growth Factor Receptor 2), though it was raised against the precursor form. Human. Predicted to react with mouse based on 100% sequence homology. Immunogen sequence has 79% homology to rat. Hamster reactivity reported in scientific literature (PMID: 23763710).
Target Species Hamster