Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, PE, Clone: 89106, R&D Systems


Manufacturer: r & d systems  FAB357P100

 View more versions of this product

Catalog No. FAB357P100

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen VEGF R2/KDR/Flk-1
Format Purified
Host Species Mouse
Applications Flow Cytometry
Immunogen &italic_begin;S. frugiperda&italic_end; insect ovarian cell line &italic_begin;Sf&italic_end; 21-derived recombinant human VEGF R2/KDR/Flk-1
Isotype IgG1
Quantity 100 Tests
Clone 89106
Cross Reactivity Human
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P35968
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3791
Test Specificity Detects human VEGF R2/KDR/Flk-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) VEGF R1, rhVEGF R3 or recombinant mouse VEGF R2 is observed.
Conjugate PE
Primary or Secondary Primary