Please login to your online account to display your discounted pricing

anti-VAV2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227078SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227078SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for this VAV2 antibody is 100% homologous in human, dog, and chimpanzee 95% in mouse, Chinese hamster, mole rat, elephant, rhesus monkey, horse and guinea pig, 89% homolgous in panda, cow, zebrafish, opossum, platypus, cichlid fish, pig, and rat.
Gene Accession No. P52735
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Immunogen A portion of amino acids 760-810 from human VAV2 was used as the immunogen for this antibody.
Regulatory Status RUO
Gene ID (Entrez) 7410
Conjugate Unlabeled
Applications Western Blotting
Quantity 0.025mg
Antigen VAV2