Please login to your online account to display your discounted pricing

anti-VAV2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227078

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227078

  • / Each
 

Description & Specifications

Specifications

Antigen VAV2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. P52735
Host Species Rabbit
Immunogen A portion of amino acids 760-810 from human VAV2 was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7410
Test Specificity The amino acid sequence used as immunogen for this VAV2 antibody is 100% homologous in human, dog, and chimpanzee 95% in mouse, Chinese hamster, mole rat, elephant, rhesus monkey, horse and guinea pig, 89% homolgous in panda, cow, zebrafish, opossum, platypus, cichlid fish, pig, and rat.
Target Species Canine
Target Species Human
Target Species Primate