Please login to your online account to display your discounted pricing

anti-VASA, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 700-724 of DDX4 was used as the immunogen.
Antigen VASA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224558SS

 
novus biologicals canada
NBP224558SS
Unlabeled 0.025mg Each for $109.00

NBP224558

 
novus biologicals canada
NBP224558
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 700-724 of DDX4 was used as the immunogen.
Antigen VASA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, cow, Xenopus, rat, horse, orangutan, zebrafish, and pig.
Gene Accession No. AF262962.1
Gene ID (Entrez) 54514
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL