Please login to your online account to display your discounted pricing

anti-Vanilloid R1/TRPV1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171774

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171774

  • / Each
 

Description & Specifications

Specifications

Antigen Vanilloid R1/TRPV1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.03mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q8NER1
Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human TRPV1 protein (within residues 725-839). [Swiss-Prot Q8NER1]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7442
Target Species Human
Target Species Murine