Please login to your online account to display your discounted pricing

anti-Vanilloid R1/TRPV1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197417

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP197417

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7442
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [UniProt O35433]
Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Concentration 1.30mg/mL
Regulatory Status RUO
Format Affinity Purified
Quantity 0.1mL
Antigen Vanilloid R1/TRPV1