Please login to your online account to display your discounted pricing

anti-Vang-like Protein 2/VANGL2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 300-350 of human Van Gogh like-2 protein was used as the immunogen.
Antigen Vang-like Protein 2/VANGL2
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224704SS

 
novus biologicals canada
NBP224704SS
Unlabeled 0.025mg Each for $109.00

NBP224704

 
novus biologicals canada
NBP224704
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 300-350 of human Van Gogh like-2 protein was used as the immunogen.
Antigen Vang-like Protein 2/VANGL2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a, CRA_b and CRA_c), chimpanzee, cow, dog, mouse and rat, 93% homologous in opossum, horse and 73% homologous in chicken and xenopus.
Gene ID (Entrez) 57216
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL