Please login to your online account to display your discounted pricing

anti-Vang-like Protein 1/VANGL1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Vang-like 1 was used as the immunogen.
Antigen Vang-like Protein 1/VANGL1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224473SS

 
novus biologicals canada
NBP224473SS
Unlabeled 0.025mg Each for $139.00

NBP224473

 
novus biologicals canada
NBP224473
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Vang-like 1 was used as the immunogen.
Antigen Vang-like Protein 1/VANGL1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2, isoform 3, isoform 4, isoform 5, isoform 6 and isoform 7) and 93% homologous in mouse.
Gene ID (Entrez) 81839
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL