Please login to your online account to display your discounted pricing

anti-Vang-like Protein 1/VANGL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224473SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224473SS

  • / Each
 

Description & Specifications

Specifications

Antigen Vang-like Protein 1/VANGL1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Vang-like 1 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 81839
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2, isoform 3, isoform 4, isoform 5, isoform 6 and isoform 7) and 93% homologous in mouse.
Target Species Human