Please login to your online account to display your discounted pricing

anti-VAMP3/Cellubrevin, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human VAMP-3 was used as the immunogen.
Antigen VAMP3/Cellubrevin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224480

 
novus biologicals canada
NBP224480
Unlabeled 0.1mg Each for $307.00

NBP224480SS

 
novus biologicals canada
NBP224480SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human VAMP-3 was used as the immunogen.
Antigen VAMP3/Cellubrevin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen reacts with Human, Mouse and Rat. Predicted to cross react with New World monkey.
Gene Accession No. Q15836
Gene ID (Entrez) 9341
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL