Please login to your online account to display your discounted pricing

anti-VAMP3/Cellubrevin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224480

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224480

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen reacts with Human, Mouse and Rat. Predicted to cross react with New World monkey.
Concentration 0.5mg/mL
Immunogen A portion of amino acids 1-50 of human VAMP-3 was used as the immunogen.
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 9341
Conjugate Unlabeled
Host Species Rabbit
Gene Accession No. Q15836
Regulatory Status RUO
Format Affinity Purified
Antigen VAMP3/Cellubrevin
Applications Western Blotting