Please login to your online account to display your discounted pricing

anti-VABP, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1-35 (LQTKQRRPSKDKIEDELE) of human ABP protein were used as the immunogen (NP_055407).
Antigen VABP
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP219020

 
novus biologicals canada
NBP219020
Unlabeled 0.1mg Each for $255.80

NBP219020SS

 
novus biologicals canada
NBP219020SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1-35 (LQTKQRRPSKDKIEDELE) of human ABP protein were used as the immunogen (NP_055407).
Antigen VABP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The Uniprot number for VABP is Q9NZI2. The amino acid sequence used as immunogen has 100% identity in human, mouse, rat, pig and bovine.
Gene Accession No. Q9NZI2
Gene ID (Entrez) 30820
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL