Please login to your online account to display your discounted pricing

anti-UVRAG, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 375-425 of human UVRAG was used as the immunogen.
Antigen UVRAG
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224482

 
novus biologicals canada
NBP224482
Unlabeled 0.1mg Each for $259.02

NBP224482SS

 
novus biologicals canada
NBP224482SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 375-425 of human UVRAG was used as the immunogen.
Antigen UVRAG
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Gene Accession No. Q9P2Y5
Gene ID (Entrez) 7405
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL