Please login to your online account to display your discounted pricing

anti-UVRAG, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224482

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224482

  • / Each
 

Description & Specifications

Specifications

Antigen UVRAG
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9P2Y5
Host Species Rabbit
Immunogen A portion of amino acids 375-425 of human UVRAG was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7405
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, chimpanzee and rat, 94% homologous in dog and 78% homologous in chicken.
Target Species Canine
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat