Please login to your online account to display your discounted pricing

anti-USP7, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1050-1100 of human USP7 protein was used as the immunogen.
Antigen USP7
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224642

 
novus biologicals canada
NBP224642
Unlabeled 0.1mg Each for $257.41

NBP224642SS

 
novus biologicals canada
NBP224642SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1050-1100 of human USP7 protein was used as the immunogen.
Antigen USP7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 93% homologous in cow, chimpanzee and monkey, 87% homologous in horse, dog and 81% homologous in mouse and rat.
Gene ID (Entrez) 7874
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL